missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, 97%

Catalog No. AAH2894206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2894206 5 g
AAH2894203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2894206 Supplier Thermo Scientific Chemicals Supplier No. H2894206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171364-80-0 | C14H19BO4 | 262.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 171364-80-0
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.11
MDL Number MFCD02179438
InChI Key REIZEQZILPXYKS-UHFFFAOYSA-N
Synonym methyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonylphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid pinacolate, 4-carbomethoxyphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid, pinacol ester, 4-methoxycarbonyl phenylboronic acid pinacol ester, methyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 4-methoxycarbonyl benzeneboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid methyl ester
PubChem CID 2773500
IUPAC Name methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 77°C to 81°C
Quantity 5 g
Formula Weight 262.11
Percent Purity 97%
Chemical Name or Material 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only