missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methoxybutyrophenone 98.0+%, TCI America™
$42.36 - $125.13
Chemical Identifiers
| CAS | 4160-51-4 |
|---|---|
| Molecular Formula | C11H14O2 |
| Molecular Weight (g/mol) | 178.23 |
| MDL Number | MFCD00027138 |
| InChI Key | JLCDSZXBELPBRD-UHFFFAOYSA-N |
| Synonym | 4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone |
| PubChem CID | 77810 |
| IUPAC Name | 1-(4-methoxyphenyl)butan-1-one |
| SMILES | CCCC(=O)C1=CC=C(OC)C=C1 |
Chemical Identifiers
| 4160-51-4 | |
| 178.23 | |
| JLCDSZXBELPBRD-UHFFFAOYSA-N | |
| 77810 | |
| CCCC(=O)C1=CC=C(OC)C=C1 |
| C11H14O2 | |
| MFCD00027138 | |
| 4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone | |
| 1-(4-methoxyphenyl)butan-1-one |
Specifications
| 4160-51-4 | |
| C11H14O2 | |
| 1 g | |
| JLCDSZXBELPBRD-UHFFFAOYSA-N | |
| 1-(4-methoxyphenyl)butan-1-one | |
| 77810 | |
| ≥98.0% (GC) | |
| 4′-Methoxybutyrophenone |
| White-Yellow | |
| MFCD00027138 | |
| 4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone | |
| CCCC(=O)C1=CC=C(OC)C=C1 | |
| 178.23 | |
| 178.23 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No