missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxybutyrophenone 98.0+%, TCI America™

Catalog No. M14361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14361G 1 g
M14365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14361G Supplier TCI America Supplier No. M14361G

Chemical Identifiers

CAS 4160-51-4
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00027138
InChI Key JLCDSZXBELPBRD-UHFFFAOYSA-N
Synonym 4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone
PubChem CID 77810
IUPAC Name 1-(4-methoxyphenyl)butan-1-one
SMILES CCCC(=O)C1=CC=C(OC)C=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Methoxybutyrophenone