missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxybutyrophenone, 97%

CAS: 4160-51-4 | C11H14O2 | 178.23 g/mol

$53.12 - $656.70

Chemical Identifiers

CAS 4160-51-4
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00027138
InChI Key JLCDSZXBELPBRD-UHFFFAOYSA-N
Synonym 4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone
PubChem CID 77810
IUPAC Name 1-(4-methoxyphenyl)butan-1-one
SMILES CCCC(=O)C1=CC=C(OC)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1173703
View Documents
Thermo Scientific Chemicals
A1173703
1 g
Each for $53.12
Only null left
 
AAA1173706
View Documents
Thermo Scientific Chemicals
A1173706
5 g
Each for $150.22
Only null left
 
AAA1173714
View Documents
Thermo Scientific Chemicals
A1173714
25 g
Each for $656.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4160-51-4
178.23
JLCDSZXBELPBRD-UHFFFAOYSA-N
77810
CCCC(=O)C1=CC=C(OC)C=C1
C11H14O2
MFCD00027138
4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone
1-(4-methoxyphenyl)butan-1-one

Specifications

4160-51-4
C11H14O2
MFCD00027138
908286
JLCDSZXBELPBRD-UHFFFAOYSA-N
1-(4-methoxyphenyl)butan-1-one
77810
97%
124°C to 128°C (4 mmHg)
1.539
1 g
4'-methoxybutyrophenone, 1-4-methoxyphenyl butan-1-one, p-methoxybutyrophenone, 4-methoxybutyrophenone, 1-butanone, 1-4-methoxyphenyl, butyrophenone, 4'-methoxy, 1-4-methoxyphenyl-1-butanone, 4-butyrylanisole, acmc-209jkw, 4'-methoxy-butyrophenone
CCCC(=O)C1=CC=C(OC)C=C1
178.23
178.23
4'-Methoxybutyrophenone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-995-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only