missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl isocyanate, 99%

Catalog No. AC319400250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319400250 25 g Glass bottle
AC319400010 1 g Glass bottle
AC319400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC319400250 Supplier Thermo Scientific Chemicals Supplier No. 319400250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56651-60-6 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56651-60-6
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00673064
InChI Key QRBHVARIMDDOOV-UHFFFAOYSA-N
Synonym 4-methoxybenzyl isocyanate, 1-isocyanatomethyl-4-methoxybenzene, p-methoxybenzyl isocyanate, 4-methoxybenzylisocyanate, isocyanic acid, p-methoxybenzyl, benzene, 1-isocyanatomethyl-4-methoxy, 4-methoxyphenyl methanisocyanate, acmc-20amf5, p-methoxybenzylisocyanate, p-metoxybenzyl-isocyanate
PubChem CID 187910
IUPAC Name 1-(isocyanatomethyl)-4-methoxybenzene
SMILES COC1=CC=C(CN=C=O)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Boiling Point 90.0°C (0.6 mmHg)
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5305 to 1.5325
Quantity 25 g
Formula Weight 163.18
Percent Purity 99%
Chemical Name or Material 4-Methoxybenzyl isocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only