missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoylacetonitrile, 98%

CAS: 3672-47-7 | C10H9NO2 | 175.187 g/mol

Supplier:  Thermo Scientific Chemicals A1597306

 View more versions of this product

Catalog No. AAA1597306

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methoxybenzoylacetonitrile
125°C to 130°C
MFCD00067889
743909
IKEPUFCALLUUBC-UHFFFAOYSA-N
3-(4-methoxyphenyl)-3-oxopropanenitrile
233398
98%
3672-47-7
C10H9NO2
5 g
3-4-methoxyphenyl-3-oxopropanenitrile, 4-methoxybenzoylacetonitrile, 4'-methoxybenzoylacetonitrile, 4-methoxybenzoyl acetonitrile, pubchem10102, 4-cyanoacetyl anisole, maybridge1_004582, acmc-1agd6, p-methoxybenzoylacetonitrile, toslab 6389
COC1=CC=C(C=C1)C(=O)CC#N
175.187
175.19

Chemical Identifiers

3672-47-7
175.187
IKEPUFCALLUUBC-UHFFFAOYSA-N
233398
COC1=CC=C(C=C1)C(=O)CC#N
C10H9NO2
MFCD00067889
3-4-methoxyphenyl-3-oxopropanenitrile, 4-methoxybenzoylacetonitrile, 4'-methoxybenzoylacetonitrile, 4-methoxybenzoyl acetonitrile, pubchem10102, 4-cyanoacetyl anisole, maybridge1_004582, acmc-1agd6, p-methoxybenzoylacetonitrile, toslab 6389
3-(4-methoxyphenyl)-3-oxopropanenitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only