missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoylacetonitrile, 98%

Catalog No. AAA1597314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1597314 25 g
AAA1597306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1597314 Supplier Thermo Scientific Chemicals Supplier No. A1597314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3672-47-7 | C10H9NO2 | 175.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3672-47-7
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00067889
InChI Key IKEPUFCALLUUBC-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-3-oxopropanenitrile, 4-methoxybenzoylacetonitrile, 4'-methoxybenzoylacetonitrile, 4-methoxybenzoyl acetonitrile, pubchem10102, 4-cyanoacetyl anisole, maybridge1_004582, acmc-1agd6, p-methoxybenzoylacetonitrile, toslab 6389
PubChem CID 233398
IUPAC Name 3-(4-methoxyphenyl)-3-oxopropanenitrile
SMILES COC1=CC=C(C=C1)C(=O)CC#N

Specifications

Melting Point 125°C to 130°C
Quantity 25 g
Beilstein 743909
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzoylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only