missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzohydrazide 98.0+%, TCI America™

Catalog No. p-7132844
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17705G 5 g
M177025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17705G Supplier TCI America Supplier No. M17705G
Only null left

Chemical Identifiers

CAS 3290-99-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017073
InChI Key REKQLYUAUXYJSZ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrazide, p-anisohydrazide, p-anisic hydrazide, p-anisoylhydrazine, 4-methoxybenzoic acid hydrazide, 4-methoxybenzoylhydrazine, anisoylhydrazine, anisic hydrazide, 4-methoxybenzene-1-carbohydrazide, p-anisic acid, hydrazide
PubChem CID 76792
IUPAC Name 4-methoxybenzohydrazide
SMILES COC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzohydrazide
RTECSNumber BZ4800000
TSCA No