missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzhydrazide, 98+%

Catalog No. p-7020904
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1301509 10 g
AAA1301518 50 g
AAA1301530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1301509 Supplier Thermo Scientific Chemicals Supplier No. A1301509

CAS: 3290-99-1 | C8H10N2O2 | 166.18 g/mol

4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3290-99-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017073
InChI Key REKQLYUAUXYJSZ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrazide, p-anisohydrazide, p-anisic hydrazide, p-anisoylhydrazine, 4-methoxybenzoic acid hydrazide, 4-methoxybenzoylhydrazine, anisoylhydrazine, anisic hydrazide, 4-methoxybenzene-1-carbohydrazide, p-anisic acid, hydrazide
PubChem CID 76792
IUPAC Name 4-methoxybenzohydrazide
SMILES COC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 136°C to 140°C
Quantity 10 g
Beilstein 389017
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzhydrazide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-952-5
RTECSNumber BZ4800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only