missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzhydrazide, 98+%
Description
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 136°C to 140°C |
| Quantity | 50 g |
| Beilstein | 389017 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 166.18 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Methoxybenzhydrazide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 221-952-5 |
| RTECSNumber | BZ4800000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only