Learn More
4-Methoxybenzenethiol, 98%, Thermo Scientific™
$143.37 - $590.80
Chemical Identifiers
| CAS | 696-63-9 |
|---|---|
| Molecular Formula | C7H8OS |
| Molecular Weight (g/mol) | 140.2 |
| MDL Number | MFCD00004849 |
| InChI Key | NIFAOMSJMGEFTQ-UHFFFAOYSA-N |
| Synonym | 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol |
| PubChem CID | 12765 |
| IUPAC Name | 4-methoxybenzenethiol |
| SMILES | COC1=CC=C(C=C1)S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC149150050
|
N/A | 5mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC149150250
|
N/A | 25mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C7H8OS | |
| MFCD00004849 | |
| 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol | |
| 4-methoxybenzenethiol |
Specifications
| 696-63-9 | |
| 100.0 | |
| 228.0°C | |
| Authentic | |
| C7H8OS | |
| CH3OC6H4SH | |
| 06, 859 | |
| 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol | |
| NIFAOMSJMGEFTQ-UHFFFAOYSA-N | |
| 4-methoxybenzenethiol | |
| 12765 | |
| 98% |
| 97.5 | |
| 1.1400g/mL | |
| 96°C | |
| 97.5% min. (GC) | |
| 1.5821 to 1.5841 | |
| MFCD00004849 | |
| 1.14 | |
| Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethylether,benzene,,chloroform | |
| COC1=CC=C(C=C1)S | |
| 140.2 | |
| 140.2 | |
| 4-Methoxybenzenethiol, 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 211-799-2
RTECSNumber : DC1800000
RUO – Research Use Only