Learn More
4-Methoxybenzenesulfonyl Chloride, 99%
CAS: 98-68-0 | C7H7ClO3S | 206.64 g/mol
$120.36 - $120.36
Chemical Identifiers
| CAS | 98-68-0 |
|---|---|
| Molecular Formula | C7H7ClO3S |
| Molecular Weight (g/mol) | 206.64 |
| MDL Number | MFCD00007446 |
| InChI Key | DTJVECUKADWGMO-UHFFFAOYSA-N |
| Synonym | 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy |
| PubChem CID | 7401 |
| IUPAC Name | 4-methoxybenzenesulfonyl chloride |
| SMILES | COC1=CC=C(C=C1)S(Cl)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC125730250
|
Thermo Scientific Chemicals
125730250 |
25 g | Glass bottle |
Each for $120.36
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 98-68-0 | |
| 206.64 | |
| DTJVECUKADWGMO-UHFFFAOYSA-N | |
| 7401 | |
| COC1=CC=C(C=C1)S(Cl)(=O)=O |
| C7H7ClO3S | |
| MFCD00007446 | |
| 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy | |
| 4-methoxybenzenesulfonyl chloride |
Specifications
| 98-68-0 | |
| 100.0 | |
| Beige to White | |
| Authentic | |
| Glass bottle | |
| CH3OC6H4SO2Cl | |
| 25 g | |
| 09,297 | |
| Solubility in water: decomposes | |
| COC1=CC=C(C=C1)S(Cl)(=O)=O | |
| 206.64 | |
| 206.65 | |
| Crystalline Solid |
| 98.5 | |
| 40.0°C to 42.0°C | |
| >110°C | |
| 98.5% min. (GC) | |
| C7H7ClO3S | |
| MFCD00007446 | |
| 11, 243 | |
| 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy | |
| DTJVECUKADWGMO-UHFFFAOYSA-N | |
| 4-methoxybenzenesulfonyl chloride | |
| 7401 | |
| 99% | |
| 4-Methoxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-692-1
RUO – Research Use Only