missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyazobenzene 98.0+%, TCI America™

Catalog No. P014225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P014225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P014225G Supplier TCI America Supplier No. P014225G
Only null left

Chemical Identifiers

CAS 2396-60-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00039804
InChI Key LGCRPKOHRIXSEG-UHFFFAOYSA-N
Synonym p-Phenylazoanisole
PubChem CID 16966
IUPAC Name (4-methoxyphenyl)-phenyldiazene
SMILES COC1=CC=C(C=C1)N=NC2=CC=CC=C2

Specifications

Melting Point 56°C
Boiling Point 340°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyazobenzene