missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyacetophenone 99.0+%, TCI America™

Catalog No. p-7131990
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M010525G 25 g
M0105500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M010525G Supplier TCI America Supplier No. M010525G
Only null left

Chemical Identifiers

CAS 100-06-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008745
InChI Key NTPLXRHDUXRPNE-UHFFFAOYSA-N
Synonym 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
PubChem CID 7476
ChEBI CHEBI:86567
IUPAC Name 1-(4-methoxyphenyl)ethan-1-one
SMILES COC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 38°C
Color White
Boiling Point 260°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Methoxyacetophenone
EINECSNumber (3)-2676
RTECSNumber AM9240000
TSCA Yes
Recommended Storage Refrigerator