missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-trans-beta-styrylboronic acid pinacol ester, 97%
CAS: 149777-83-3 | C15H21BO3 | 260.14 g/mol
Supplier: Thermo Scientific Chemicals H3037306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Methoxy-trans-beta-styrylboronic acid pinacol ester | |
| C15H21BO3 | |
| 5 g | |
| VXSHTICWQKLRMP-UHFFFAOYSA-N | |
| 2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 15419573 | |
| 98% |
| 149777-83-3 | |
| MFCD04038966 | |
| 4-methoxy-trans-beta-styrylboronic acid pinacol ester, e-2-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl, 2-e-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-methoxy-trans-beta-styrylboronicacidpinacolester, 2-4-methoxyphenyl-vinylboronic acid pinacol ester, 2-e-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-2-4-methoxyphenyl vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| COC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1 | |
| 260.14 | |
| 260.14 |
Chemical Identifiers
| 149777-83-3 | |
| 260.14 | |
| VXSHTICWQKLRMP-UHFFFAOYSA-N | |
| 15419573 | |
| COC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1 |
| C15H21BO3 | |
| MFCD04038966 | |
| 4-methoxy-trans-beta-styrylboronic acid pinacol ester, e-2-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl, 2-e-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-methoxy-trans-beta-styrylboronicacidpinacolester, 2-4-methoxyphenyl-vinylboronic acid pinacol ester, 2-e-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-2-4-methoxyphenyl vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only