missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-trans-beta-styrylboronic acid pinacol ester, 97%

Catalog No. AAH30373MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH30373MD 250 mg
AAH3037303 1 g
AAH3037306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH30373MD Supplier Thermo Scientific Chemicals Supplier No. H30373MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149777-83-3 | C15H21BO3 | 260.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149777-83-3
Molecular Formula C15H21BO3
Molecular Weight (g/mol) 260.14
MDL Number MFCD04038966
InChI Key VXSHTICWQKLRMP-UHFFFAOYSA-N
Synonym 4-methoxy-trans-beta-styrylboronic acid pinacol ester, e-2-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl, 2-e-2-4-methoxyphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-methoxy-trans-beta-styrylboronicacidpinacolester, 2-4-methoxyphenyl-vinylboronic acid pinacol ester, 2-e-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-methoxystyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-2-4-methoxyphenyl vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 15419573
IUPAC Name 2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES COC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1

Specifications

Quantity 250 mg
Formula Weight 260.14
Percent Purity 98%
Chemical Name or Material 4-Methoxy-trans-beta-styrylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only