missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-4'-methyldiphenylamine 98.0+%, TCI America™
Supplier: TCI America M207025G
Specifications
| 4-Methoxy-4′-methyldiphenylamine | |
| 84°C | |
| MFCD00092922 | |
| N-(4-Methoxyphenyl)-p-tolylamine | |
| COC1=CC=C(NC2=CC=C(C)C=C2)C=C1 | |
| 213.28 | |
| 213.28 | |
| Crystalline Powder |
| 39253-43-5 | |
| C14H15NO | |
| 25 g | |
| KIDXWDVZFZMXGM-UHFFFAOYSA-N | |
| N-(4-methoxyphenyl)-4-methylaniline | |
| 11435828 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 39253-43-5 | |
| 213.28 | |
| KIDXWDVZFZMXGM-UHFFFAOYSA-N | |
| 11435828 | |
| COC1=CC=C(NC2=CC=C(C)C=C2)C=C1 |
| C14H15NO | |
| MFCD00092922 | |
| N-(4-Methoxyphenyl)-p-tolylamine | |
| N-(4-methoxyphenyl)-4-methylaniline |
Safety and Handling
TSCA : No