missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-pyridineboronic acid hydrate, 97%

Catalog No. p-3212125
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378382500 250 mg Glass bottle
AC378380010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378382500 Supplier Thermo Scientific Chemicals Supplier No. 378382500

CAS: 1266615-79-5 | C6H8BNO3 | 152.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1266615-79-5 , 355004-67-0
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD07186213
InChI Key YUTPAZKVEOJQCY-UHFFFAOYSA-N
Synonym 4-methoxy-3-pyridineboronic acid, 4-methoxypyridin-3-yl boronic acid, 4-methoxypyridine-3-boronic acid, 3-borono-4-methoxypyridine, 4-methoxy-3-pyridylboronic acid, boronic acid, 4-methoxy-3-pyridinyl, 4-methoxy-pyridine-3-boric acid, acmc-209iha, 4-methoxy-5-pyridineboronic acid, 4-methoxy-pyridin-3-boronic acid
PubChem CID 2762556
IUPAC Name (4-methoxypyridin-3-yl)boronic acid
SMILES COC1=CC=NC=C1B(O)O

Specifications

Color Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 152.94
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methoxy-3-pyridineboronic acid hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only