missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-buten-2-one, tech. 90%

Catalog No. AAA1123509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1123509 10 g
AAA1123518 50 g
AAA1123530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1123509 Supplier Thermo Scientific Chemicals Supplier No. A1123509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4652-27-1 | C5H8O2 | 100.12 g/mol

4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 4652-27-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008778,MFCD00008778,MFCD00008778
InChI Key VLLHEPHWWIDUSS-ARJAWSKDSA-N
Synonym z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one
PubChem CID 643837
IUPAC Name (Z)-4-methoxybut-3-en-2-one
SMILES CO\C=C/C(C)=O

Specifications

Density 0.99
Boiling Point 66°C to 69°C (12 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.465
Quantity 10 g
Beilstein 1741053
Solubility Information Miscible with tetrahydrofuran,ether,acetonitrile and benzene.
Formula Weight 100.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Methoxy-3-buten-2-one
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 225-087-4
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only