missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3,5-dimethyl-2-pyridinemethanol 98.0+%, TCI America™
Supplier: TCI America M13275G
Specifications
| 4-Methoxy-3,5-dimethyl-2-pyridinemethanol | |
| 61°C | |
| MFCD01076195 | |
| 2-Hydroxymethyl-4-methoxy-3,5-dimethylpyridine | |
| COC1=C(C)C=NC(CO)=C1C | |
| 167.21 | |
| 167.21 | |
| Crystalline Powder |
| 86604-78-6 | |
| C9H13NO2 | |
| 5 g | |
| PSEPRWKZZJWRCB-UHFFFAOYSA-N | |
| (4-methoxy-3,5-dimethylpyridin-2-yl)methanol | |
| 737823 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 86604-78-6 | |
| 167.21 | |
| PSEPRWKZZJWRCB-UHFFFAOYSA-N | |
| 737823 | |
| COC1=C(C)C=NC(CO)=C1C |
| C9H13NO2 | |
| MFCD01076195 | |
| 2-Hydroxymethyl-4-methoxy-3,5-dimethylpyridine | |
| (4-methoxy-3,5-dimethylpyridin-2-yl)methanol |
Safety and Handling
TSCA : No