missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. M18851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18851G 1 g
M18855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18851G Supplier TCI America Supplier No. M18851G

Chemical Identifiers

CAS 556812-41-0
Molecular Formula C11H15F3O4SSi
Molecular Weight (g/mol) 328.377
InChI Key IPZYLSPWZUDOHB-UHFFFAOYSA-N
Synonym 4-Methoxy-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 4-Methoxy-2-(trimethylsilyl)phenyl Ester
PubChem CID 12182644
IUPAC Name (4-methoxy-2-trimethylsilylphenyl) trifluoromethanesulfonate
SMILES COC1=CC(=C(C=C1)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 328.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate