missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. m22551g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22551G Supplier TCI America Supplier No. M22551G

Chemical Identifiers

CAS 208399-66-0
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.98
MDL Number MFCD02684315
InChI Key AMSQNQJCBXQYEX-UHFFFAOYSA-N
PubChem CID 2773486
IUPAC Name (4-methoxy-2-methylphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(C)=C1

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 1 g
Synonym 4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6
Formula Weight 165.98
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride)