missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 52289-54-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD02261771
InChI Key WICYVKGMEJSDAO-UHFFFAOYSA-N
Synonym 2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
PubChem CID 283285
IUPAC Name 4-methoxy-2-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)OC)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427020010
SDS View Documents
Thermo Scientific Chemicals
427020010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427020050
SDS View Documents
Thermo Scientific Chemicals
427020050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

52289-54-0
150.18
WICYVKGMEJSDAO-UHFFFAOYSA-N
283285
CC1=C(C=CC(=C1)OC)C=O
C9H10O2
MFCD02261771
2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
4-methoxy-2-methylbenzaldehyde

Specifications

52289-54-0
100.0
262°C to 267°C
Authentic
Glass bottle
CH30C6H3(CH3)CHO
1g
2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
CC1=C(C=CC(=C1)OC)C=O
150.18
150.18
4-Methoxy-2-methylbenzaldehyde
96.0
1.1070g/mL
>110°C
96% min. (GC)
C9H10O2
MFCD02261771
1.107
WICYVKGMEJSDAO-UHFFFAOYSA-N
4-methoxy-2-methylbenzaldehyde
283285
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only