missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2(5H)-furanone, 97%

Catalog No. p-3758302
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276680050 5 g Glass bottle
AC276680250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC276680050 Supplier Thermo Scientific Chemicals Supplier No. 276680050

CAS: 69556-70-3 | C5H6O3 | 114.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69556-70-3
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00071565
InChI Key VOYDEHILKLSVNN-UHFFFAOYSA-N
PubChem CID 643477
IUPAC Name 3-methoxy-2H-furan-5-one
SMILES COC1=CC(=O)OC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Yellow
Boiling Point 130°C to 131°C (18.0 mbar)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Synonym 4-methoxy-2 5h-furanone, 4-methoxyfuran-2 5h-one, 4-methoxy-5h-furan-2-one, 2 5h-furanone,4-methoxy, 2 5h-furanone, 4-methoxy, 4-methoxy-5-hydrofuran-2-one
Solubility Information Solubility in water: 240g/L (20°C)
Formula Weight 114.1
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxy-2(5H)-furanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only