missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. p-7132591
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
M11971G 1 g
M11975G 5 g
2 options

Catalog No. M11971G

Supplier: TCI America M11971G

Only null left

Chemical Identifiers

CAS 95407-69-5
Molecular Formula C10H20NO2
MDL Number MFCD00270334
Synonym 4-Methoxy-TEMPO

Specifications

Melting Point 43°C
Quantity 1 g
Formula Weight 186.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical [Catalyst for Oxidation]