missing translation for 'onlineSavingsMsg'
Learn More

4-Mercapto-4-methyl-2-pentanone, 98%

Catalog No. p-7030825
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3169303 1 g
AAH3169306 5 g
AAH3169314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3169303 Supplier Thermo Scientific Chemicals Supplier No. H3169303
Only null left

CAS: 19872-52-7 | C6H12OS | 132.221 g/mol

4-Mercapto-4-methyl-2-pentanone is one of the most strongly contributing odorants in the volatile fraction of a Japanese green tea (sen-cha) infusion, and on the basis of the results of an aroma extract dilution analysis. 4-mercapto-4-methyl-2-pentanone in the grapefruit/sulfury group was found in reconstituted concentrate after thermal concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Mercapto-4-methyl-2-pentanone is one of the most strongly contributing odorants in the volatile fraction of a Japanese green tea (sen-cha) infusion, and on the basis of the results of an aroma extract dilution analysis. 4-mercapto-4-methyl-2-pentanone in the grapefruit/sulfury group was found in reconstituted concentrate after thermal concentration.

Solubility
Soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 19872-52-7
Molecular Formula C6H12OS
Molecular Weight (g/mol) 132.221
MDL Number MFCD00085208
InChI Key QRNZMFDCKKEPSX-UHFFFAOYSA-N
Synonym 4-mercapto-4-methyl-2-pentanone, 4-mercapto-4-methylpentan-2-one, 2-pentanone, 4-mercapto-4-methyl, 4-methyl-4-thiolpentan-2-one, 4-methyl-4-mercapto-2-pentanone, 4-methyl-4-mercaptopentan-2-one, unii-9524rg5zql, 4-methyl-4-sulfanyl-2-pentanone, 4-sulfanyl-4-methylpentan-2-one, 4-sulphanyl-4-methylpentan-2-one
PubChem CID 88290
ChEBI CHEBI:77856
IUPAC Name 4-methyl-4-sulfanylpentan-2-one
SMILES CC(=O)CC(C)(C)S

Specifications

Density 0.961
Boiling Point 174°C
Flash Point 54°C (129°F)
Odor Unpleasant
Refractive Index 1.462
Quantity 1 g
UN Number UN3336
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material 4-Mercapto-4-methyl-2-pentanone
Show More Show Less
Hazard Category H226-H319-H335
Hazard Statement GHS H Statement
H226-H319-H335
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 243-386-8
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only