missing translation for 'onlineSavingsMsg'
Learn More

4-(m-Tolylamino)pyridine-3-sulfonamide 98.0+%, TCI America™

Catalog No. p-7137526
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30075G Supplier TCI America Supplier No. T30075G
Only null left

Chemical Identifiers

CAS 72811-73-5
Molecular Formula C12H13N3O2S
Molecular Weight (g/mol) 263.315
MDL Number MFCD00661332
InChI Key ZXPCUGWAKUIOOF-UHFFFAOYSA-N
Synonym 4-(3-Methylphenyl)aminopyridine-3-sulfonamide, 4-(m-Tolylamino)-3-sulfamoylpyridine, 4-(3-Methylphenyl)amino-3-sulfamoylpyridine
PubChem CID 819349
IUPAC Name 4-(3-methylanilino)pyridine-3-sulfonamide
SMILES CC1=CC(=CC=C1)NC2=C(C=NC=C2)S(=O)(=O)N

Specifications

Melting Point 170°C
Color White
Quantity 5 g
Formula Weight 263.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(m-Tolylamino)pyridine-3-sulfonamide
TSCA No
Recommended Storage Refrigerator