missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ketoisophorone 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 222°C |
| Quantity | 25 g |
| Formula Weight | 152.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 4-Ketoisophorone |
Safety and Handling
| EINECSNumber | (3)-4529 |
| TSCA | Yes |