missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylpiperidine 98.0+%, TCI America™

Catalog No. I07335G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07335G 5 g
I07331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07335G Supplier TCI America Supplier No. I07335G
Only null left

Chemical Identifiers

CAS 19678-58-1
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD09055342
InChI Key YBPWIUSXQXYTSR-UHFFFAOYSA-N
PubChem CID 140569
IUPAC Name 4-(propan-2-yl)piperidine
SMILES CC(C)C1CCNCC1

Specifications

Color Yellow
Boiling Point 171°C
Quantity 5 g
UN Number 1993
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropylpiperidine