missing translation for 'onlineSavingsMsg'
Learn More

4-(Isopropylcarbamoyl)benzeneboronic acid, 95%

Catalog No. p-7035211
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH52925MD 250 mg
AAH5292503 1 g
2 options

Catalog No. AAH52925MD

Supplier: Thermo Scientific Chemicals H52925MD

Only null left

CAS: 397843-67-3 | C10H14BNO3 | 207.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 397843-67-3
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.036
MDL Number MFCD04039353
InChI Key GBCSEYKTZAKRMT-UHFFFAOYSA-N
Synonym 4-isopropylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenylboronic acid, 4-isopropylcarbamoyl benzeneboronic acid, 4-isopropylcarbamoyl phenylboronic acid, 4-propan-2-ylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenyl boronic acid, n-isopropyl 4-boronobenzamide, boronic acid, 4-1-methylethyl amino carbonyl phenyl, 4-1-methylethyl amino carbonyl phenyl-boronic acid
PubChem CID 2773475
IUPAC Name [4-(propan-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC(C)C)(O)O

Specifications

Melting Point 162°C to 167°C
Quantity 250 mg
Formula Weight 207.04
Percent Purity 95%
Chemical Name or Material 4-(Isopropylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only