missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbiphenyl, TCI America™

Catalog No. p-7131252
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I01715G 5 g
I017125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I01715G Supplier TCI America Supplier No. I01715G
Only null left

Chemical Identifiers

CAS 7116-95-2
Molecular Formula C15H16
Molecular Weight (g/mol) 196.29
MDL Number MFCD00221718
InChI Key KWSHGRJUSUJPQD-UHFFFAOYSA-N
Synonym 4-isopropylbiphenyl, p-isopropyldiphenyl, biphenyl, 4-isopropyl, 1,1'-biphenyl, 4-1-methylethyl, unii-4kbv29724i, 4-phenylcumene, acmc-209ohg, 4,4'-isopropylbiphenyl, 4-isopropyl-1,1'-biphenyl, 4-isopropyl-1,1'-biphenyl #
PubChem CID 23503
IUPAC Name 4-(propan-2-yl)-1,1'-biphenyl
SMILES CC(C)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 291°C
Quantity 5 g
Formula Weight 196.29
Physical Form Liquid
Chemical Name or Material 4-Isopropylbiphenyl
EINECSNumber (4)-0797
RTECSNumber DV5415000
TSCA No