missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropylbenzyl Alcohol 98.0+%, TCI America™
$49.36 - $117.72
Chemical Identifiers
| CAS | 536-60-7 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.221 |
| MDL Number | MFCD00004663 |
| InChI Key | OIGWAXDAPKFNCQ-UHFFFAOYSA-N |
| Synonym | 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol |
| PubChem CID | 325 |
| ChEBI | CHEBI:27628 |
| IUPAC Name | (4-propan-2-ylphenyl)methanol |
| SMILES | CC(C)C1=CC=C(C=C1)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I027225ML
|
TCI America
I027225ML |
25 mL |
Each for $49.36
|
|
|||||
|
I0272100ML
|
TCI America
I0272100ML |
100 mL |
Each for $117.72
|
|
|||||
Chemical Identifiers
| 536-60-7 | |
| 150.221 | |
| OIGWAXDAPKFNCQ-UHFFFAOYSA-N | |
| 325 | |
| (4-propan-2-ylphenyl)methanol |
| C10H14O | |
| MFCD00004663 | |
| 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol | |
| CHEBI:27628 | |
| CC(C)C1=CC=C(C=C1)CO |
Specifications
| 536-60-7 | |
| 248°C | |
| MFCD00004663 | |
| 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol | |
| CC(C)C1=CC=C(C=C1)CO | |
| 150.221 | |
| CHEBI:27628 | |
| ≥98.0% (GC) | |
| 4-Isopropylbenzyl Alcohol |
| Yellow | |
| C10H14O | |
| 25 mL | |
| OIGWAXDAPKFNCQ-UHFFFAOYSA-N | |
| (4-propan-2-ylphenyl)methanol | |
| 325 | |
| 150.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1049
RTECSNumber : GZ7260000
TSCA : Yes