missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. I027225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I027225ML 25 mL
I0272100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I027225ML Supplier TCI America Supplier No. I027225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 536-60-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00004663
InChI Key OIGWAXDAPKFNCQ-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl alcohol, cuminol, cumic alcohol, cuminyl alcohol, cuminic alcohol, 4-isopropylphenyl methanol, cumyl alcohol, p-cymen-7-ol, cumin alcohol, p-isopropylbenzyl alcohol
PubChem CID 325
ChEBI CHEBI:27628
IUPAC Name (4-propan-2-ylphenyl)methanol
SMILES CC(C)C1=CC=C(C=C1)CO

Specifications

Color Yellow
Boiling Point 248°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzyl Alcohol
EINECSNumber (3)-1049
RTECSNumber GZ7260000
TSCA Yes