missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzoic Acid 98.0+%, TCI America™

Catalog No. I016925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I016925G Supplier TCI America Supplier No. I016925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 536-66-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002564
InChI Key CKMXAIVXVKGGFM-UHFFFAOYSA-N
Synonym 4-isopropylbenzoic acid, cuminic acid, cumic acid, p-isopropylbenzoic acid, 4-1-methylethyl benzoic acid, benzoic acid, 4-1-methylethyl, 4-propan-2-yl benzoic acid, benzoic acid, p-isopropyl, p-cumic acid, 4-isopropyl benzoic acid
PubChem CID 10820
ChEBI CHEBI:28122
IUPAC Name 4-propan-2-ylbenzoic acid
SMILES CC(C)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropylbenzoic Acid
EINECSNumber (3)-1338
RTECSNumber DH3850500
TSCA Yes