missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzoic acid, 98+%

Catalog No. p-3758270
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412800050 5 g Glass bottle
AC412800250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412800050 Supplier Thermo Scientific Chemicals Supplier No. 412800050

CAS: 536-66-3 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 536-66-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00002564
InChI Key CKMXAIVXVKGGFM-UHFFFAOYSA-N
Synonym 4-isopropylbenzoic acid, cuminic acid, cumic acid, p-isopropylbenzoic acid, 4-1-methylethyl benzoic acid, benzoic acid, 4-1-methylethyl, 4-propan-2-yl benzoic acid, benzoic acid, p-isopropyl, p-cumic acid, 4-isopropyl benzoic acid
PubChem CID 10820
ChEBI CHEBI:28122
IUPAC Name 4-propan-2-ylbenzoic acid
SMILES CC(C)C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 116.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4COOH
Quantity 5 g
Merck Index 15, 2608
Solubility Information Solubility in water: sparingly soluble
Formula Weight 164.2
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-642-5

RUO – Research Use Only