missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzaldehyde, tech. 90%

Catalog No. AAA1401914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1401914 25 g
AAA1401922 100 g
AAA1401936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1401914 Supplier Thermo Scientific Chemicals Supplier No. A1401914

CAS: 122-03-2 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-03-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00006953
InChI Key WTWBUQJHJGUZCY-UHFFFAOYSA-N
Synonym 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
PubChem CID 326
ChEBI CHEBI:28671
SMILES CC(C)C1=CC=C(C=O)C=C1

Specifications

Density 0.979
Boiling Point 234°C to 235°C
Flash Point 93°C (199°F)
Refractive Index 1.53
Quantity 25 g
Beilstein 636547
Sensitivity Air Sensitive
Merck Index 14,2621
IUPAC Name 4-(propan-2-yl)benzaldehyde
Formula Weight 148.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Isopropylbenzaldehyde
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 204-516-9
RTECSNumber CU7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only