missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropylantipyrine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 104°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 230.31 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Isopropylantipyrine |
Safety and Handling
| EINECSNumber | (9)-0193 |
| RTECSNumber | CD2800000 |
| TSCA | Yes |