missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Isopropylamino)butanol 98.0+%, TCI America™
$119.30 - $413.83
Chemical Identifiers
| CAS | 42042-71-7 |
|---|---|
| Molecular Formula | C7H17NO |
| Molecular Weight (g/mol) | 131.219 |
| InChI Key | IPLWOCGPIGUXOR-UHFFFAOYSA-N |
| PubChem CID | 12950986 |
| IUPAC Name | 4-(propan-2-ylamino)butan-1-ol |
| SMILES | CC(C)NCCCCO |
Chemical Identifiers
| 42042-71-7 | |
| 131.219 | |
| 12950986 | |
| CC(C)NCCCCO |
| C7H17NO | |
| IPLWOCGPIGUXOR-UHFFFAOYSA-N | |
| 4-(propan-2-ylamino)butan-1-ol |
Specifications
| 42042-71-7 | |
| 85°C | |
| 1 g | |
| IPLWOCGPIGUXOR-UHFFFAOYSA-N | |
| 4-(propan-2-ylamino)butan-1-ol | |
| 12950986 | |
| ≥98.0% (GC,T) | |
| 4-(Isopropylamino)butanol |
| Colorless | |
| C7H17NO | |
| 1993 | |
| CC(C)NCCCCO | |
| 131.219 | |
| 131.22 | |
| Liquid |
Safety and Handling
TSCA : No