missing translation for 'onlineSavingsMsg'
Learn More

4'-Isopropylacetophenone 95.0+%, TCI America™

Catalog No. I05705ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05705ML 5 mL
I057025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05705ML Supplier TCI America Supplier No. I05705ML

Chemical Identifiers

CAS 645-13-6
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00048297
InChI Key PDLCCNYKIIUWHA-UHFFFAOYSA-N
Synonym 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one
PubChem CID 12578
IUPAC Name 1-[4-(propan-2-yl)phenyl]ethan-1-one
SMILES CC(C)C1=CC=C(C=C1)C(C)=O

Specifications

Color Yellow
Boiling Point 238°C
Quantity 5 mL
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Isopropylacetophenone