missing translation for 'onlineSavingsMsg'
Learn More

4'-Isopropylacetophenone, 97%

Catalog No. AAB2058122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2058122 100 g
AAB2058106 5 g
AAB2058114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2058122 Supplier Thermo Scientific Chemicals Supplier No. B2058122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 645-13-6 | C11H14O | 162.23 g/mol

4'-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 645-13-6
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00048297
InChI Key PDLCCNYKIIUWHA-UHFFFAOYSA-N
Synonym 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one
PubChem CID 12578
IUPAC Name 1-(4-propan-2-ylphenyl)ethanone
SMILES CC(C)C1=CC=C(C=C1)C(C)=O

Specifications

Density 0.97
Boiling Point 238°C
Refractive Index 1.523
Quantity 100 g
Beilstein 2205694
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 4'-Isopropylacetophenone
EINECSNumber 211-433-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only