missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropoxybenzaldehyde 96.0+%, TCI America™
$98.18 - $340.75
Chemical Identifiers
| CAS | 18962-05-5 |
|---|---|
| Molecular Formula | C10H12O2 |
| Molecular Weight (g/mol) | 164.204 |
| MDL Number | MFCD00052357 |
| InChI Key | WDANSDASCKBVKH-UHFFFAOYSA-N |
| Synonym | 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde |
| PubChem CID | 250077 |
| IUPAC Name | 4-propan-2-yloxybenzaldehyde |
| SMILES | CC(C)OC1=CC=C(C=C1)C=O |
Chemical Identifiers
| 18962-05-5 | |
| 164.204 | |
| WDANSDASCKBVKH-UHFFFAOYSA-N | |
| 250077 | |
| CC(C)OC1=CC=C(C=C1)C=O |
| C10H12O2 | |
| MFCD00052357 | |
| 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde | |
| 4-propan-2-yloxybenzaldehyde |
Specifications
| 18962-05-5 | |
| 112°C | |
| MFCD00052357 | |
| 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde | |
| CC(C)OC1=CC=C(C=C1)C=O | |
| 164.204 | |
| 164.20 | |
| Liquid |
| Yellow | |
| C10H12O2 | |
| 5 g | |
| WDANSDASCKBVKH-UHFFFAOYSA-N | |
| 4-propan-2-yloxybenzaldehyde | |
| 250077 | |
| ≥96.0% (GC) | |
| 4-Isopropoxybenzaldehyde |
Safety and Handling
TSCA : No