missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzaldehyde, 97%

CAS: 18962-05-5 | C10H12O2 | 164.204 g/mol

$66.37 - $736.14

Chemical Identifiers

CAS 18962-05-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00052357
InChI Key WDANSDASCKBVKH-UHFFFAOYSA-N
Synonym 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde
PubChem CID 250077
IUPAC Name 4-propan-2-yloxybenzaldehyde
SMILES CC(C)OC1=CC=C(C=C1)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1276003
View Documents
Thermo Scientific Chemicals
A1276003
1 g
Each for $66.37
Only null left
 
AAA1276006
View Documents
Thermo Scientific Chemicals
A1276006
5 g
Each for $183.59
Only null left
 
AAA1276014
View Documents
Thermo Scientific Chemicals
A1276014
25 g
Each for $736.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18962-05-5
164.204
WDANSDASCKBVKH-UHFFFAOYSA-N
250077
CC(C)OC1=CC=C(C=C1)C=O
C10H12O2
MFCD00052357
4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde
4-propan-2-yloxybenzaldehyde

Specifications

18962-05-5
C10H12O2
MFCD00052357
742921
4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde
CC(C)OC1=CC=C(C=C1)C=O
164.204
164.21
4-Isopropoxybenzaldehyde
108°C to 110°C (5 mmHg)
1.547
1 g
Air Sensitive
WDANSDASCKBVKH-UHFFFAOYSA-N
4-propan-2-yloxybenzaldehyde
250077
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only