missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzaldehyde, 97%

Catalog No. AAA1276014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276014 25 g
AAA1276003 1 g
AAA1276006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1276014 Supplier Thermo Scientific Chemicals Supplier No. A1276014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18962-05-5 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18962-05-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00052357
InChI Key WDANSDASCKBVKH-UHFFFAOYSA-N
Synonym 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde
PubChem CID 250077
IUPAC Name 4-propan-2-yloxybenzaldehyde
SMILES CC(C)OC1=CC=C(C=C1)C=O

Specifications

Boiling Point 108°C to 110°C (5 mmHg)
Refractive Index 1.547
Quantity 25 g
Beilstein 742921
Sensitivity Air Sensitive
Formula Weight 164.21
Percent Purity 97%
Chemical Name or Material 4-Isopropoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only