missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-4-thien-2-yltetrahydropyran, 95%, Thermo Scientific™

Chemical Identifiers

CAS 926921-63-3
Molecular Formula C10H11NO2S
Molecular Weight (g/mol) 209.26
MDL Number MFCD09879931
InChI Key CDJWJYYYKMFRFC-UHFFFAOYSA-N
Synonym 4-isocyanato-4-thiophen-2-yl oxane, 4-isocyanato-4-thien-2-yl tetrahydro-2h-pyran, 4-thien-2-yltetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thien-2-yltetrahydropyran, 2-4-isocyanatotetrahydro-2h-pyran-4-yl thiophene, 4-thien-2-yl tetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thiophen-2-yltetrahydropyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
PubChem CID 24229667
IUPAC Name 4-isocyanato-4-thiophen-2-yloxane
SMILES O=C=NC1(CCOCC1)C1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC62206CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62206DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

926921-63-3
209.26
CDJWJYYYKMFRFC-UHFFFAOYSA-N
24229667
O=C=NC1(CCOCC1)C1=CC=CS1
C10H11NO2S
MFCD09879931
4-isocyanato-4-thiophen-2-yl oxane, 4-isocyanato-4-thien-2-yl tetrahydro-2h-pyran, 4-thien-2-yltetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thien-2-yltetrahydropyran, 2-4-isocyanatotetrahydro-2h-pyran-4-yl thiophene, 4-thien-2-yl tetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thiophen-2-yltetrahydropyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
4-isocyanato-4-thiophen-2-yloxane

Specifications

926921-63-3
100.0
C10H11NO2S
4-isocyanato-4-thiophen-2-yl oxane, 4-isocyanato-4-thien-2-yl tetrahydro-2h-pyran, 4-thien-2-yltetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thien-2-yltetrahydropyran, 2-4-isocyanatotetrahydro-2h-pyran-4-yl thiophene, 4-thien-2-yl tetrahydro-2h-pyran-4-yl isocyanate, 4-isocyanato-4-thiophen-2-yltetrahydropyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
O=C=NC1(CCOCC1)C1=CC=CS1
209.26
209.27
4-Isocyanato-4-thien-2-yltetrahydropyran
94.5
95%
MFCD09879931
CDJWJYYYKMFRFC-UHFFFAOYSA-N
4-isocyanato-4-thiophen-2-yloxane
24229667
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents