missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isobutylmorpholine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 167°C |
| Quantity | 25 mL |
| UN Number | 1993 |
| Formula Weight | 143.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Isobutylmorpholine |
Safety and Handling
| EINECSNumber | (5)-0860 |
| TSCA | No |