missing translation for 'onlineSavingsMsg'
Learn More

4'-Isobutylacetophenone, 97%

Catalog No. AAA1062430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062430 250 g
AAA1062418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1062430 Supplier Thermo Scientific Chemicals Supplier No. A1062430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38861-78-8 | C12H16O | 176.259 g/mol

4'-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.

Solubility
Miscible with chloroform and methanol. Slightly miscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with reducing agents, strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 38861-78-8
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00027393
InChI Key KEAGRYYGYWZVPC-UHFFFAOYSA-N
Synonym 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
PubChem CID 93214
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanone
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C

Specifications

Density 0.952
Flash Point 54°C (129°F)
Refractive Index 1.518
Quantity 250 g
UN Number UN1224
Beilstein 1935275
Solubility Information Miscible with chloroform and methanol. Slightly miscible with water.
Formula Weight 176.27
Percent Purity 97%
Chemical Name or Material 4'-Isobutylacetophenone
Hazard Category H226-H317
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P272-P280-P303+P361+P353-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 254-159-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only