missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotriphenylamine 98.0+%, TCI America™

Catalog No. I09511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09511G 1 g
I09515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09511G Supplier TCI America Supplier No. I09511G
Only null left

Chemical Identifiers

CAS 38257-52-2
Molecular Formula C18H14IN
Molecular Weight (g/mol) 371.221
MDL Number MFCD11046352
InChI Key OWWVTWHBNAWUJO-UHFFFAOYSA-N
Synonym 4-Iodo-N,N-diphenylaniline
PubChem CID 18615936
IUPAC Name 4-iodo-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)I

Specifications

Melting Point 109°C
Quantity 1 g
Formula Weight 371.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodotriphenylamine