missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotetrahydro-2H-pyran, ≥97%, Thermo Scientific™

Catalog No. CC29911DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29911DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29911DA Supplier Thermo Scientific Chemicals Supplier No. CC29911DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25637-18-7
Molecular Formula C5H9IO
Molecular Weight (g/mol) 212.03
MDL Number MFCD06797467
InChI Key JTRNQTFTRDPITG-UHFFFAOYSA-N
Synonym 4-iodotetrahydro-2h-pyran, 4-iodo-tetrahydro-2h-pyran, 4-iodotetrahydropyran, 4-iodo-tetrahydropyran, 4-iodo-tetrahydro-pyran, 2h-pyran, tetrahydro-4-iodo, tetrahydro-4-iodo-2h-pyran, 4-iodanyloxane, 4-iodooxacyclohexane, 4-iodopentahydropyran
PubChem CID 2795506
IUPAC Name 4-iodooxane
SMILES IC1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 212.03
Percent Purity 95%
Chemical Name or Material 4-Iodotetrahydro-2H-pyran
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.