missing translation for 'onlineSavingsMsg'
Learn More

4-Iodopyridine, 96%

Catalog No. AAB2545906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2545906 5 g
AAB2545903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2545906 Supplier Thermo Scientific Chemicals Supplier No. B2545906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15854-87-2 | C5H4IN | 205.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15854-87-2
Molecular Formula C5H4IN
Molecular Weight (g/mol) 205.00
MDL Number MFCD02093937
InChI Key RTLUPHDWSUGAOS-UHFFFAOYSA-N
Synonym pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
PubChem CID 609492
IUPAC Name 4-iodopyridine
SMILES IC1=CC=NC=C1

Specifications

Melting Point 96°C to 99°C
Quantity 5 g
Beilstein 1594
Sensitivity Light sensitive
Formula Weight 205
Percent Purity 96%
Chemical Name or Material 4-Iodopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only