missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodopyrazole 98.0+%, TCI America™
$76.43 - $346.71
Chemical Identifiers
| CAS | 3469-69-0 |
|---|---|
| Molecular Formula | C3H3IN2 |
| Molecular Weight (g/mol) | 193.975 |
| MDL Number | MFCD00005244 |
| InChI Key | LLNQWPTUJJYTTE-UHFFFAOYSA-N |
| Synonym | 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295 |
| PubChem CID | 77022 |
| ChEBI | CHEBI:45260 |
| IUPAC Name | 4-iodo-1H-pyrazole |
| SMILES | C1=C(C=NN1)I |
Chemical Identifiers
| 3469-69-0 | |
| 193.975 | |
| LLNQWPTUJJYTTE-UHFFFAOYSA-N | |
| 77022 | |
| 4-iodo-1H-pyrazole |
| C3H3IN2 | |
| MFCD00005244 | |
| 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295 | |
| CHEBI:45260 | |
| C1=C(C=NN1)I |
Specifications
| 3469-69-0 | |
| White-Yellow | |
| MFCD00005244 | |
| 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295 | |
| C1=C(C=NN1)I | |
| 193.975 | |
| CHEBI:45260 | |
| ≥98.0% (GC,T) | |
| 4-Iodopyrazole |
| 109°C | |
| C3H3IN2 | |
| 5 g | |
| LLNQWPTUJJYTTE-UHFFFAOYSA-N | |
| 4-iodo-1H-pyrazole | |
| 77022 | |
| 193.98 | |
| Crystalline Powder |
Safety and Handling
TSCA : No